ODN 2006 (Type B) (synthetisch), TLRgrade™
Lieferant: ENZO LIFE SCIENCES
ALX746056M001
ENZOALX746056M001EA
841
EUR
ENZOALX746056M001
ODN 2006 (Type B) (synthetisch), TLRgrade™
ODN 2006 (Type B)
CpG oligodeoxynucleotide (Type B) with phosphorothioate backbone. Specific ligand for human and mouse TLR9 (Toll-like receptor 9).
- TLR9 ligand
Lieferumfang: Includes 1 vial of double distilled water (endotoxin-free).
Spezifikation Testergebnisse
Source | Synthetic |
Formulation | Lyophilised. Sterile. |
Purity | Activity and endotoxin tested - TLRgrade™ |
Endotoxin content | <0.002 EU/µg |
Quantity | 128.9 nmol |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...