ODN 2395 (synthetisch), TLRgrade™

Lieferant: ENZO LIFE SCIENCES

ALX746070M001
ENZOALX746070M001EA 841 EUR
ENZOALX746070M001
ODN 2395 (synthetisch), TLRgrade™
ODN 2395

TLR9 ligand.


CpG oligodeoxynucleotide (Type C) with phosphorothioate backbone. Specific ligand for human and mouse TLR9 (Toll-like receptor 9).


Lieferumfang: Includes 1 vial of double distilled water (endotoxin-free).

Bestellen

Spezifikation Testergebnisse

Source Synthetic
Formulation Lyophilised. Sterile.
Purity Activity and endotoxin tested - TLRgrade™
Endotoxin content <0.002 EU/µg
Quantity 143 nmol (~1000 µg)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR