ODN 2118 (Control for ODN 1585) (synthetisch), TLRgrade™
Lieferant: ENZO LIFE SCIENCES
ALX746203C100
ENZOALX746203C100EA
195
EUR
ENZOALX746203C100
ODN 2118 (Control for ODN 1585) (synthetisch), TLRgrade™
ODN 2118 (Control for ODN 1585)
TLR9 activator.
GpC oligodeoxynucleotide with mixed phosphodiester/phosphorothioate backbone.
Lieferumfang: Includes 1 vial of double distilled water (endotoxin-free).
Spezifikation Testergebnisse
Source | Synthetic |
Formulation | Lyophilised. Sterile. |
Purity | Activity and endotoxin tested - TLRgrade™ |
Endotoxin content | <0.002 EU/µg |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...