Duo-iODN (inhibitory ODN) (synthetisch) endotoxinfrei
Lieferant: ENZO LIFE SCIENCES
ALX746255C100
ENZOALX746255C100EA
190
EUR
ENZOALX746255C100
Duo-iODN (inhibitory ODN) (synthetisch) endotoxinfrei
Duo-iODN (inhibitory ODN)
Inhibitory iODN oligodeoxynucleotide with phosphorothioate backbone. Sequence of an inhibitory ODN for in vivo use in rodents (50-150µg per injection): prototype class I that potently inhibits TLR9 and TLR7 signaling.
- Inhibitor of TLR7 and TLR9