Neutral-ODN (control for iODNs)

Lieferant: ENZO LIFE SCIENCES

ALX746256C100
ENZOALX746256C100EA 204 EUR
ENZOALX746256C100
Neutral-ODN (control for iODNs)
Neutral-ODN (control for iODNs)

Inactive (neutral) control.


Inactive (neutral) control ODN for in vivo use in rodents (50 – 150 µg per injection) without agonistic nor antagonistic activity.


Working concentration depends upon concentration, type and MW of stimulating CpG ODN, the cellular system and read-out used.


Reconstitution:
For a 100µM stock solution, dissolve the total vial content in 270µl endotoxin-free water (included) or PBS (to order separately).To obtain optimal dissolving we recommend the following procedure:
- Add 50% of the solvent and let dissolve for 10 min.
- Add remaining 50% of the solvent and mix thoroughly.
- Moderate warming may aid dissolving.


Lieferumfang: Includes 1 vial of double distilled water (endotoxin-free).

Bestellen

Spezifikation Testergebnisse

Source Synthetic
Formulation Lyophilised. Sterile.
Endotoxin content <0.002 EU/µg
Quantity ~20 nmol (100 µg)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR