Duo-iODN (inhibitory ODN) (synthetisch) endotoxinfrei (Bulk)
Lieferant: ENZO LIFE SCIENCES
ALX746355M001
ENZOALX746355M001EA
570
EUR
ENZOALX746355M001
Duo-iODN (inhibitory ODN) (synthetisch) endotoxinfrei (Bulk)
Duo-iODN (inhibitory ODN)
Inhibitory iODN oligodeoxynucleotide with phosphorothioate backbone. Sequence of an inhibitory ODN for in vivo use in rodents (50-150µg per injection): prototype class I that potently inhibits TLR9 and TLR7 signaling.
- Inhibitor of TLR7 and TLR9