Cu/Zn-Superoxide dismutase ELISA kit
Lieferant: ENZO LIFE SCIENCES
ALX850033KI01
ENZOALX850033KI01EA 1090 EUR
ENZOALX850033KI01
Cu/Zn-Superoxide dismutase ELISA kit
Tests ELISAs
For the quantitative determination of human Cu/Zn SOD in cell culture supernatants, serum and other body fluids.
  • Assay Time: <2 hours
Late stages of apoptosis are characterized by the loss of membrane integrity and a release of the intracellular Cu/Zn SOD.