Annexin V (Human) ELISA Kit
Lieferant: ENZO LIFE SCIENCES
ALX850049KI01
ENZOALX850049KI01EA
1050
EUR
ENZOALX850049KI01
Annexin V (Human) ELISA Kit
Tests
ELISAs
For the quantitative detemination of human annexin V in cell culture supernatant and serum.
- Assay Time: 3,5 hours