Annexin V (Human) ELISA Kit
Lieferant: ENZO LIFE SCIENCES
ALX850049KI01
ENZOALX850049KI01EA 1050 EUR
ENZOALX850049KI01
Annexin V (Human) ELISA Kit
Tests ELISAs
For the quantitative detemination of human annexin V in cell culture supernatant and serum.
  • Assay Time: 3,5 hours