Caspase-3 fluorometric assay kit
Lieferant: ENZO LIFE SCIENCES
ALX850216KI01
ENZOALX850216KI01EA 426 EUR
ENZOALX850216KI01
Caspase-3 fluorometric assay kit
Tests Enzymtests
Based on the detection of AFC (7-amino-4-trifluoromethyl coumarin) after cleavage from the labelled substrate DEVD-AFC.
Lieferumfang: Cell Lysis Buffer (25 ml), 2X Reaction Buffer (2 ml), 1 mM DEVD-AFC (125 µl), 1 M DTT (100 µl).