Caspase inhibitor set plus
Lieferant: ENZO LIFE SCIENCES
Synthetic peptide inhibitors that irreversibly inhibit the activity of each of the caspase-family proteases. YVAD is the recognition sequence for caspase-1, VDVAD is the recognition sequence for caspase-2, DEVD is the recognition sequence for caspase-3, VEID is the recognition sequence for caspase-6, IETD is the recognition sequence for caspase-8, LEHD is the recognition sequence for caspase-9, and VAD is the recognition sequence for a caspase(s) that functions upstream of most other caspases.
Bestellinformation: Contents: Contains 25 µl (2mM) each of the following inhibitors:
- Caspase-1 Inhibitor, Z-YVAD-FMK, yellow cap
- Caspase-2 Inhibitor, Z-VDVAD-FMK, orange cap
- Caspase-3 Inhibitor, Z-DEVD-FMK, red cap
- Caspase-6 Inhibitor, Z-VEID-FMK, violet cap
- Caspase-8 Inhibitor, Z-IETD-FMK, green cap
- Caspase-9 Inhibitor, Z-LEHD-FMK, blue cap
- Caspase-Family Inhibitor, Z-VAD-FMK, amber cap