Recombinant Caspase (active) set lll
Lieferant: ENZO LIFE SCIENCES
ALX850243KI01
ENZOALX850243KI01EA 1660 EUR
ENZOALX850243KI01
Recombinant Caspase (active) set lll
Enzyme
We recommend using 1 U per assay for analyzing caspase activity.
Active recombinant caspases are useful in screening caspase inhibitors, studying enzyme kinetics and regulation, determining substrate specificity, and as a positive control in caspase activity assays. 
Bestellinformation: Contents: Contains 25 units each of the following caspases:
- Caspase-1 (active) (human) (recombinant), yellow cap
- Caspase-2 (active) (human) (recombinant), orange cap
- Caspase-3 (active) (human) (recombinant), red cap
- Caspase-6 (active) (human) (recombinant), violet cap
- Caspase-7 (active) (human) (recombinant), red cap
- Caspase-8 (active) (human) (recombinant), green cap
- Caspase-9 (active) (human) (recombinant), blue cap
- Caspase-10 (active) (human) (recombinant), amber cap