CD40 Soluble (Human) ELISA kit
Lieferant: ENZO LIFE SCIENCES
ALX850262KI01
ENZOALX850262KI01EA
1030
EUR
ENZOALX850262KI01
CD40 Soluble (Human) ELISA kit
Tests
ELISAs
For the quantitative determination of human soluble CD40 in cell culture supernatants, serum and other body fluids.
- Assay Time: <3,25 hours