Mitochondria/cytosol fractionation kit
Lieferant: ENZO LIFE SCIENCES
ALX850276KI01
ENZOALX850276KI01EA 725 EUR
ENZOALX850276KI01
Mitochondria/cytosol fractionation kit
Tests Zelltests
For the effective isolation of a highly enriched mitochondrial fraction from the cytosolic fraction of various mammalian cells including both apoptotic an non-apoptotic cells. The enriched mitochondrial and cytosolic fractions can be used to study apoptosis and signal trasduction pathways by detecting translocations of factors of interest between the two fractions by ELISA, Western blot or other assays.
The core kit contains unique formulations of buffers and reagents that allow isolation of mitochondria and cytosol in only two easy steps. No ultracentrifugations are required. No toxic chemicals are involved.
Lieferumfang: Kit includes Mitochondria Extraction Buffer (10 ml), 5X Cytosol Extraction Buffer (20 ml), 1 M DTT (110 µl), Protease Inhibitor Cocktail (1 vial).