ADMA (human) ELISA kit
Lieferant: ENZO LIFE SCIENCES
ALX850323KI01
ENZOALX850323KI01EA 1880 EUR
ENZOALX850323KI01
ADMA (human) ELISA kit
Tests ELISAs
For the quantitative determination of asymmetrical dimethylarginine (ADMA) in human serum, citrate- and EDTA-plasma. Does not cross-react with SDMA, NMMA or L-arginine.