ADMA (human) ELISA kit
Lieferant: ENZO LIFE SCIENCES
ALX850323KI01
ENZOALX850323KI01EA
1880
EUR
ENZOALX850323KI01
ADMA (human) ELISA kit
Tests
ELISAs
For the quantitative determination of asymmetrical dimethylarginine (ADMA) in human serum, citrate- and EDTA-plasma. Does not cross-react with SDMA, NMMA or L-arginine.
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)