ADMA direct (mouse/rat) ELISA kit
Lieferant: ENZO LIFE SCIENCES
ALX850327KI01
ENZOALX850327KI01EA 2060 EUR
ENZOALX850327KI01
ADMA direct (mouse/rat) ELISA kit
Tests ELISAs
For the quantitative detection of asymmetrical dimethylarginine (ADMA) in mouse or rat EDTA-plasma and serum. Does not cross-react with SDMA, NMMA or L-arginine.