ADMA direct (mouse/rat) ELISA kit
Lieferant: ENZO LIFE SCIENCES
ALX850327KI01
ENZOALX850327KI01EA
2060
EUR
ENZOALX850327KI01
ADMA direct (mouse/rat) ELISA kit
Tests
ELISAs
For the quantitative detection of asymmetrical dimethylarginine (ADMA) in mouse or rat EDTA-plasma and serum. Does not cross-react with SDMA, NMMA or L-arginine.
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)