Ion channel ligand library, Screen-Well®
Lieferant: ENZO LIFE SCIENCES
BML28050100 BML28050500
ENZOBML28050100EA 951 EUR
ENZOBML28050100 ENZOBML28050500
Ion channel ligand library, Screen-Well®
Zellkultur-Ergänzungen und -Zusätze
The Screen-Well® ion channel ligand library contains 71 ion channel blockers and openers for use in characterising and identifying ion channels in individual cells or tissue.