Dopaminergic ligand library, Screen-Well®
Lieferant: ENZO LIFE SCIENCES
The Screen-Well® dopaminergic ligands plate contains 80 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs. The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The dopaminergic ligands plate is 1 plate of the larger neurotransmitter library which contains over 650 ligands.