Cholinergic ligand library, Screen-Well®
Lieferant: ENZO LIFE SCIENCES
BML28150100
BML28150500
ENZOBML28150100EA
1320
EUR
ENZOBML28150100
ENZOBML28150500
Cholinergic ligand library, Screen-Well®
Zellkultur-Ergänzungen und -Zusätze
The cholinergic ligands plate contains 72 compounds ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs. The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The cholinergic ligands plate is 1 plate of the larger neurotransmitter library which contains over 650 ligands.