Metabotropic glutamatergic ligand library, Screen-Well®
Lieferant: ENZO LIFE SCIENCES
BML28180100 BML28180500
ENZOBML28180100EA 1330 EUR
ENZOBML28180100 ENZOBML28180500
Metabotropic glutamatergic ligand library, Screen-Well®
Zellkultur-Ergänzungen und -Zusätze
The Screen-Well® metabotropic glutamatergic ligands plate contains 49 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs. The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The metabotropic glutamatergic ligands plate is 1 plate of the larger neurotransmitter library which contains over 650 ligands.