Kinase inhibitor library, Screen-Well®
Lieferant: ENZO LIFE SCIENCES
The Screen-Well™ Kinase Inhibitor Library contains 80 known kinase inhibitors of well-defined activity. Inhibitors are supplied dissolved in DMSO at 10 mM and aliquoted into deep-well plates at either 100 or 500 ml per well. The library is an ideal tool for chemical genomics, assay development and other pharmacological applications. Includes inhibitors of these important kinases: Insulin/IGF Receptors, PI 3-Kinase, CaM Kinase II, JAK, PKA, CDK, JNK, PKC, CKI II, MAPK, RAF, EGFR, MEK, SAPK, GSK, MLCK, Src-family, IKK, PDGFR, VEGFR and many more.
Bestellinformation: Quantity: 100 µl or 500 µl per well. Contents: 80 compounds including inhibitors of these important kinases: BTK, CaM Kinase, CDK, CKI & II, EGFR, GSK, IKK, Insulin receptor, JAK, JNK, MAPK, MEK, MLCK, PI 3-Kinase, PDGFR, PKA, PKC, RAF, SAPK, Src-family, VEGFR, and more.