Epigenetics library, Screen-Well®
Lieferant: ENZO LIFE SCIENCES
BML28360100 BML28360500
ENZOBML28360100EA 1590 EUR
ENZOBML28360100 ENZOBML28360500
Epigenetics library, Screen-Well®
Zellkultur-Ergänzungen und -Zusätze
The Screen-Well® epigenetics library contains 43 compounds with defined activity against enzymes which carry out epigenetic modification of lysine. It also includes DNA methylation inhibitors. Compounds are dissolved in DMSO at 10 mM and aliquoted into deep-well plates at 100 µl or 500 µl per well. The library is a useful tool for chemical genomics, assay development and other pharmacological applications. The library contains inhibitors of these important enzymes: HDACs, SIRTs, Lysine demethylases, HATs, Histone methyl transferases, DNA methyltransferases as well as SIRT activators. A variety of structurally and mechanistically different compound classes are included.
The Screen-Well® Epigenetics library contains 43 compounds with defined activity against enzymes which carry out epigenetic modification of lysine. It also includes DNA methylation inhibitors. Compounds are dissolved in DMSO at 10mM and aliquoted into deep-well plates at 100 or 500µL per well. The library is a useful tool for chemical genomics, assay development and other pharmacological applications. The library contains inhibitors of these important enzymes: HDACs, SIRTs, Lysine demethylases, HATs, Histone methyl transferases, DNA methyltransferases as well as SIRT activators. A variety of structurally and mechanistically different compound classes are included.
Bestellinformation: Contains 43 compounds supplied in DMSO at a concentration of 10mM.