Hepatotoxicity library, Screen-Well®
Lieferant: ENZO LIFE SCIENCES
BML28510100 BML28510500
ENZOBML28510100EA 4000 EUR
ENZOBML28510100 ENZOBML28510500
Hepatotoxicity library, Screen-Well®
Zellkultur-Ergänzungen und -Zusätze
The Screen-Well® hepatotoxicity library is a focused collection of 238 compounds with defined and diverse hepatotoxicity, including steatosis, mitochondrial toxicity, Mallory body formation, cholestatic effects, and many more. A variety of structurally and mechanistically different compound classes are included, as well as nontoxic controls. The compounds are dissolved in DMSO at 10 mM and aliquoted into deep-well plates. The library is an essential tool for predictive toxicology screening and assay development.
Bestellinformation: Contains 238 compounds with defined and diverse hepatotoxicity. A variety of structurally and mechanistically different compound classes, as well as nontoxic controls, are included.