Clindamycin hydrochloride ≥95% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: Cleocin Hydrochloride, (2S,4R)-N-((1S,2S)-2-Chloro-1-((2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(methylthio)tetrahydro-2H-pyran-2-yl)propyl)-1-methyl-4-propylpyrrolidine-2-carboxamide hydrochloride, 7[S]-Chloro-7-deoxylincomycin, Clindamycin HCl

BMLA1900100
ENZOBMLA1900100EA 229 EUR
ENZOBMLA1900100
Clindamycin hydrochloride ≥95% (durch HPLC)
Clindamycin hydrochloride

Active against Gram-positive bacteria. Clindamycin binds to the 50S subunit of bacterial ribosomes and suppresses protein synthesis.


  • White solid


Soluble in water


Verpackung: Shipping temperature: Ambient

Formel: C₁₈H₃₃ClN₂O₅S·HCl
Molecular Weight: 461,45 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07793327
CAS-Nummer: 21462-39-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR