Daptomycin ≥93% (durch HPLC)

Lieferant: Enzo Life Sciences

BMLA2010020 BMLA2010100
ENZOBMLA2010020EA 114 EUR
ENZOBMLA2010020 ENZOBMLA2010100
Daptomycin ≥93% (durch HPLC)
Daptomycin

Lipopeptide antibiotic


Novel lipopeptide antibiotic active against Gram positive bacteria.

Formel: C₇₂H₁₀₁N₁₇O₂₆
MDL: MFCD08282794
CAS-Nummer: 103060-53-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR