Mycophenolsäure ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: (E)-6-(4-Hydroxy-6-methoxy-7-methyl-3-oxo-1,3-dihydroisobenzofuran-5-yl)-4-methylhex-4-enoic acid

BMLA2490500
ENZOBMLA2490500EA 370 EUR
ENZOBMLA2490500
Mycophenolsäure ≥98% (durch TLC)
Mycophenolsäure

Inosine-5'-monophosphate dehydrogenase inhibitor


Antiviral and immunosuppressive activity. Clinically useful antipsoriatic agent. Mycophenolic acid prodrugs are effective in rheumatoid arthritis and for the prevention of organ transplant rejection. Mechanism of action is inhibition of inosine-5'-monophosphate dehydrogenase.


Isolated from Penicillium brevi-compactum

Formel: C₁₇H₂₀O₆
Molecular Weight: 320.34 g/mol
MDL: MFCD00036814
CAS-Nummer: 24280-93-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR