Spectinomycin sulphate ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

BMLA2810010
ENZOBMLA2810010EA 86.9 EUR
ENZOBMLA2810010
Spectinomycin sulphate ≥98% (durch HPLC)
Spectinomycin sulphate

Antibiotic. Inhibits protein synthesis (elongation) by interfering with peptidyl tRNA translocation. Effective against Gram-negative and Gram-positive bacteria (Gonnococcus only). Mutation in rpsE (the gene for ribosomal protein S5) prevents binding of spectinomycin.


  • White solid


Soluble in water


Verpackung: Shipping temperature: Ambient

Formel: C₁₄H₂₄N₂O₇·H₂SO₄
Lagertemperatur: Gefrierschrank
MDL: MFCD00270185
CAS-Nummer: 23312-56-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR