CV-Cathepsin K detection kit
Lieferant: ENZO LIFE SCIENCES
BMLAK1260001
ENZOBMLAK1260001EA 367 EUR
ENZOBMLAK1260001
CV-Cathepsin K detection kit
Tests Zelltests
Based on the dye cresyl violet and the cathepsin K LeuArg peptide recognition sequence, this kit allows the detection of cathepsin K activity in living cells by fluorescence microscopy.
Once cells are grown, staining is completed in just a few minutes. Hoechst 33342 stain and Acridine Orange are also included to allow visualization of nuclei and lysosomes.
Elevated cathepsin enzyme activity often indicates the presence of a pathological condition including cancer, Alzheimer's disease, autoimmune disease, and osteoporosis.
Bestellinformation: Kit contains CV-(LR)2 reagent, Acridine Orange, Hoechst 33342 stain.