CV-Cathepsin L detection kit
Lieferant: ENZO LIFE SCIENCES
Based on the dye cresyl violet and the cathepsin L PheArg peptide recognition sequence, this kit allows the detection of cathepsin L activity in living cells by fluorescence microscopy.
Once cells are grown, staining is completed in just a few minutes. Hoechst 33342 stain and Acridine Orange are also included to allow visualization of nuclei and lysosomes.
Elevated cathepsin enzyme activity often indicates the presence of a pathological condition including cancer, Alzheimer's disease, autoimmune disease, and osteoporosis.
Bestellinformation: Kit contains CV-(FR)2 reagent, Acridine Orange, Hoechst 33342 stain.