Caspase-1 assay kit for drug discovery
Lieferant: ENZO LIFE SCIENCES
BMLAK7010001
ENZOBMLAK7010001EA
697
EUR
ENZOBMLAK7010001
Caspase-1 assay kit for drug discovery
Tests
Enzymtests
This kit is a complete assay system to measure protease activity of recombinant caspase-1. Cleavage of a tetrapeptide substrate is monitored colorimetrically at 405 nm or fluorimetrically (Ex. 360 nm/Em. 460 nm).
Lieferumfang: Kit includes Caspase-1 enzyme (human) (recombinant), Ac-YVAD-pNA colorimetric substrate, Ac-YVAD-AMC fluorogenic substrate, calibration standards, aldehyde inhibitor, assay buffer, half volume 96-microwell plate, and instruction booklet.