Caspase-3 cell assay kit
Lieferant: ENZO LIFE SCIENCES
This kit is a complete assay system for measuring caspase-3 and caspase-3-like activity (DEVDase) in cellular extracts.
It is performed in a 96-well format with all reagents for preparing cell extracts, measuring caspase activity and calibrating the assay. The caspase-3, is useful as a positive control, for testing cellular extracts for endogenous inhibitors, and more
Lieferumfang: Kit includes Caspase-3 enzyme (human, recombinant), pNA Substrate, pNA calibration standard, AMC Substrate, AMC calibration standard (7-amino-4-methylcoumarin), Inhibitor (Ac-DEvD-CHO), Assay buffer, Cell lysis buffer, Half volume microplate.