Cyclo [Arg-Ala-Asp-D-Phe-Val]
Lieferant: ENZO LIFE SCIENCES
BMLAM1010001
ENZOBMLAM1010001EA 114 EUR
ENZOBMLAM1010001
Cyclo [Arg-Ala-Asp-D-Phe-Val]
Proteine und Peptide
Negative control for integrin avß3 antagonist Cyclo [Arg-Gly-Asp-D-Phe-Val].