Sulindac Sulphide ≥99% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

BMLAP2000005 BMLAP2000025
ENZOBMLAP2000005EA 84.7 EUR
ENZOBMLAP2000005 ENZOBMLAP2000025
Sulindac Sulphide ≥99% (durch TLC)
Sulindac Sulphide

Active metabolite of sulindac. Inhibits cyclooxygenase. More selective for PGHS-1 (ID50=0.5 µM) over PGHS-2 (ID50=14 µM). Induces apoptosis by a cyclooxygenase-independent mechanism. Inhibits colorectal cancer growth and γ-secretase. NF-κB inhibitor. Inhibits Ras signaling.


  • COX inhibitor


Yellow-orange solid.


Soluble in DMSO (>25mg/ml) or 100% ethanol (5mg/ml, warm).

Formel: C₂₀H₁₇FO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00869764
CAS-Nummer: 32004-67-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR