3-Methyladenin ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: 3-Methyl-3H-adenin, 6-Amino-3-methylpurin

BMLAP5020025
ENZOBMLAP5020025EA 102 EUR
ENZOBMLAP5020025
3-Methyladenin ≥98% (durch HPLC)
3-Methyladenin

PI3 kinase inhibitor


PI3 kinase inhibitor and a well-known inhibitor of autophagy. This compound has been shown to induce autophagy under nutrient-rich conditions as well as inhibiting starvation-induced autophagy due to differential effects on class I versus class III PI3 kinase.

Formel: C₆H₇N₅
Molecular Weight: 149,16 g/mol
MDL: MFCD00010531
CAS-Nummer: 5142-23-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR