(±)-Cyanopindolol hemifumarate salt ≥99% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

BMLAR1010050
ENZOBMLAR1010050EA 371 EUR
ENZOBMLAR1010050
(±)-Cyanopindolol hemifumarate salt ≥99% (durch TLC)
(±)-Cyanopindolol hemifumarate salt

Adrenergic receptor and serotonin receptor antagonist.


ß-Adrenergic receptor antagonist. Serotonin 5HT1A/1B antagonist.


Soluble in warm water (2 mg/ml), DMSO (8 mg/ml).

Formel: C₁₆H₂₁N₃O₂
MDL: MFCD28100500
CAS-Nummer: 69906-85-0

Bestellen

Spezifikation Testergebnisse

Purity ≥99% (TLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR