Ac-Ala-Pro-Nle-Asp-H
Lieferant: ENZO LIFE SCIENCES
BMLAW94850100
ENZOBMLAW94850100EA 159 EUR
ENZOBMLAW94850100
Ac-Ala-Pro-Nle-Asp-H
Proteine und Peptide
Highly specific aldehyde based inhibitor of the caspase-like peptidic activity of the 20S proteasome.