Ac-Arg-Leu-Arg-AMC
Lieferant: ENZO LIFE SCIENCES
BMLAW97850005
ENZOBMLAW97850005EA 373 EUR
ENZOBMLAW97850005
Ac-Arg-Leu-Arg-AMC
Proteine und Peptide
This fluorogenic peptide is often the substrate of choice for assaying the trypsin-like activity of purified proteasomes. Benefits include a low Km, high specific activity, and the fact that the substrate is cleaved exclusively at the R-AMC bond.