Calcimycin (A23187) (aus Streptomyces chartreusis) ≥99% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: Calcium Ionophore A23187, Calcium Ionophore III, Antibiotic A23187

BMLCA1000010
ENZOBMLCA1000010EA 172 EUR
ENZOBMLCA1000010
Calcimycin (A23187) (aus Streptomyces chartreusis) ≥99% (durch TLC)
Calcimycin (A23187)

Antibiotic possessing weak in vitro antimicrobial activity against gram positive bacteria and fungi. A highly selective divalent cation ionophore, commonly used to increase intracellular Ca2⁺ levels in intact cells.


Off-white solid


Soluble in DMSO, 100% ethanol or methylene chloride (10 mg/mL); slightly soluble in water. Solubilize in DMSO or ethanol prior to preparing aqueous solutions of very low concentrations.


Verpackung: Shipping temperature: Ambient

Formel: C₂₉H₃₇N₃O₆
Molecular Weight: 523,63 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00151202
CAS-Nummer: 52665-69-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR