Niguldipine hydrochloride ≥98%
Lieferant: ENZO LIFE SCIENCES
Ca²⁺ channel blocker
A classic L-type Ca²⁺ channel blocker (IC50=75nM) which can also inhibit T-type Ca²⁺ channels (IC50=244nM). It is a novel inhibitor of drug transport by p-glycoprotein (both stereoisomers are equipotent at inhibition of p-glycoprotein).
CAS-Nummer:
119934-51-9 |