N-(6-Aminohexyl)-1-naphthalinsulfonamidhydrochlorid ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: W-5 Hydrochloride

BMLCA3150020
ENZOBMLCA3150020EA 180 EUR
ENZOBMLCA3150020
N-(6-Aminohexyl)-1-naphthalinsulfonamidhydrochlorid ≥98% (durch HPLC)
N-(6-Aminohexyl)-1-naphthalinsulfonamidhydrochlorid

Binds to calmodulin, inhibiting Ca²⁺-calmodulin-regulated enzyme activity. Cell permeable.


  • Calmodulin inhibitor


White to off-white solid.


Soluble in water, DMSO (20 mg/ml), 100% ethanol (hot) or methanol (hot).

Formel: C₁₆H₂₂N₂O₂S
Molecular Weight: 342,88 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00058043
CAS-Nummer: 61714-25-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR