N-(6-Aminohexyl)-5-chlor-1-naphthalinsulfonamidhydrochlorid ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: W-7 Hydrochloride, N-(6-Aminohexyl)-5-chloronaphthalene-1-sulphonamide hydrochloride

BMLCA3200050
ENZOBMLCA3200050EA 131 EUR
ENZOBMLCA3200050
N-(6-Aminohexyl)-5-chlor-1-naphthalinsulfonamidhydrochlorid ≥98% (durch TLC)
N-(6-Aminohexyl)-5-chlor-1-naphthalinsulfonamidhydrochlorid

Binds to calmodulin, inhibiting Ca²⁺-calmodulin-regulated enzyme activity. It inhibits the Ca²⁺/calmodulin-induced activation of myosin light chain kinase in a concentration dependent manner (IC50=25 µM). Cell permeable.


  • Calmodulin inhibitor


White to off-white solid.


Soluble in water (warm), methanol, 100% ethanol (10 mg/ml) (warm), DMSO (25 mg/ml), dimethyl formamide or ethylene glycol.

Formel: C₁₆H₂₁ClN₂O₂S
Molecular Weight: 377,32 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00012559
CAS-Nummer: 61714-27-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR