Fluphenazine-N-2-chloroethane 2HCl 98
Lieferant: ENZO LIFE SCIENCES
Synonyme:
SKF-7171A
BMLCA3250050
ENZOBMLCA3250050EA
131
EUR
ENZOBMLCA3250050
Fluphenazine-N-2-chloroethane 2HCl 98
Fluphenazine-N-2-chloroethane 2HCl
A potent irreversible inactivator of calmodulin. Possesses potent antiproliferative and cytotoxic activity. It is also an irreversible D2 dopamine receptor antagonist.
- Calmodulin inhibitor
Pale yellow solid.
Soluble in DMSO (50mg/ml) or water (10 mg/ml warm).
Formel:
C₂₂H₂₅CIF₃N₃S·2HCl Molecular Weight: 528.6 g/mol Lagertemperatur: Raumtemperatur |
CAS-Nummer:
83016-35-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...