8-Bromo-cADP-ribose ≥95%

Lieferant: ENZO LIFE SCIENCES
Achtung

BMLCA4170100
ENZOBMLCA4170100EA 91.3 EUR
ENZOBMLCA4170100
8-Bromo-cADP-ribose ≥95%
8-Bromo-cADP-ribose

cADPR antagonist.


cADPR antagonist. Blocks the cADPR-induced release of Ca²⁺ from sea urchin egg homogenates at 1 - 3.5 µM.


Soluble in water (5 mg/ml).

Formel: C₁₅H₂₀BrN₅O₁₃P₂
Molecular Weight: 620,2 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 151898-26-9

Bestellen

Spezifikation Testergebnisse

Purity ≥95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR