Nigericin sodium salt ≥98%
Lieferant: ENZO LIFE SCIENCES
Synonyme:
Sodium (R)-2-((2R,3S,6R)-6-(((2S,4R,5R,7R,9R,10R)-2-((2S,2'R,3'S,5R,5'R)-5'-((2S,3S,5R,6R)-6-hydroxy-6-(hydroxymethyl)-3,5-dimethyltetrahydro-2H-pyran-2-yl)-2,3'-dimethyloctahydro-[2,2'-bifuran]-5-yl)-9-methoxy-2,4,10-trimethyl-1,6-dioxaspiro[4.5]decan-7-yl)methyl)-3-methyltetrahydro-2H-pyran-2-yl)propanoate
NALP3 activator
Acts as an H+, K+, Na+, Pb2+ ionophore. Is most commonly an antiporter of H+ and K+. Stimulates Ca²⁺ release from mitochondrial stores by disruption of membrane potantial. Inhibits the Golgi functions. Activator of NALP3.
Formel:
C₄₀H₆₇NaO₁₁ Molecular Weight: 746,94 g/mol |
MDL:
MFCD00036825 CAS-Nummer: 28643-80-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...