Aphidicolin ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

BMLCC1010001
ENZOBMLCC1010001EA 114 EUR
ENZOBMLCC1010001
Aphidicolin ≥98% (durch TLC)
Aphidicolin

Reversible inhibitor of DNA polymerase a and d


A diterpene fungal metabolite that acts as reversible inhibitor of eukaryotic nuclear DNA replication. Useful for cell synchronization. Blocks the cell cycle at early S phase. Prolongs the half life of DNA methyltransferase. Specific inhibitor of DNA polymerase α and δ in eukaryotic cells and in some viruses of animal origin. Acts synergistically with vincristine and doxorubicin. Apoptosis inhibitor/inducer.

Formel: C₂₀H₃₄O₄
MDL: MFCD00083214
CAS-Nummer: 38966-21-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR