L-Mimosine ≥98%

Lieferant: ENZO LIFE SCIENCES

Synonyme: (S)-2-Amino-3-(3-hydroxy-4-oxopyridin-1(4H)-yl)propanoic acid

BMLCC1020500
ENZOBMLCC1020500EA 356 EUR
ENZOBMLCC1020500
L-Mimosine ≥98%
L-Mimosine

A rare plant amino acid that reversibly blocks the cell cycle at G1-phase. Inhibits the hydroxylation of deoxyhypusine and thus inhibits eukaryote initiation factor 5-A (eIF5-A). Inhibitor of mammalian tyrosinase and dopamine β-hydroxylase. Also chelates iron via the kojic acid metal-binding domain. Arrests DNA synthesis by inhibiting deoxyribonucleotide metabolism.


  • Cell cycle blocker


Pink to brown powder.


Soluble in 10% sodium bicarbonate (15mg/ml)

Formel: C₈H₁₀N₂O₄
Molecular Weight: 198,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00075909
CAS-Nummer: 500-44-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR