Fipronil ≥98%
Lieferant: ENZO LIFE SCIENCES
|
Gefahr
|
Synonyme:
5-Amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-3-cyano-4-(trifluoromethyl)sulphinylpyrazole, 5-Amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-(trifluoromethyl)sulphinylpyrazole-3-carbonitrile
BMLCM1020100
ENZOBMLCM1020100EA
299
EUR
ENZOBMLCM1020100
Fipronil ≥98%
Fipronil
Cl⁻ channel blocker.
GABA-gated chloride channel blocker. Blocks GABA responses on Xenopus oocytes. Blocks GABA-gated chloride ion channel in Drosophila.
Formel:
C₁₂H₄Cl₂F₆N₄OS Molecular Weight: 437,14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00867812 CAS-Nummer: 120068-37-3 EINECS: 424-610-5 UN: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Purity | ≥98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...