Bafilamycin A1 ≥95% (durch HPLC, TLC)

Lieferant: ENZO LIFE SCIENCES

BMLCM1100100
ENZOBMLCM1100100EA 197 EUR
ENZOBMLCM1100100
Bafilamycin A1 ≥95% (durch HPLC, TLC)
Bafilamycin A1

Macrolide antibiotic that acts as a potent and selective inhibitor of vacuolar-type H+-ATPase (V-type) (IC50=004 - 0.4 nmol/mg). Valuable tool for distinguishing among different types of ATPases. Inhibitor of endosomal acidification. Blocks lysosomal cholesterol trafficking in macrophages. Blocks pH regulation in brain cells.


  • White powder to colorless film


Soluble in 100% ethanol, methanol or DMSO

Formel: C₃₅H₅₈O₉
Lagertemperatur: Gefrierschrank
MDL: MFCD06795130
CAS-Nummer: 88899-55-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9fc2853ae79a6853e5aa822ecfddd9974ed4b284&vid=9c87045a3565f3652b722260d153d7483226695f&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR