Bongkrekic acid triammonium salt ≥92% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
BMLCM1131000
ENZOBMLCM1131000EA
1490
EUR
ENZOBMLCM1131000
Bongkrekic acid triammonium salt ≥92% (durch HPLC)
Bongkrekic acid triammonium salt
Bongkrekic acid is an inhibitory ligand of the mitochondrial adenine nucleotide translocase. It inhibits mitochondrial permeability transition pore opening. At 50 µM it inhibits dexamethasone- apoptosis in thymocytes, preventing the PARP cleavage and DEVDase (caspase-3 like) activity normally produced by these treatments.
- MPTP inhibitor
Beige to pink lyophilized solid.
Lagertemperatur:
Gefrierschrank |
CAS-Nummer:
11076-19-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...