3-Azaspiro[5.5]undecane hydrochloride ≥97% (durch HPLC)
Lieferant: Enzo Life Sciences
A novel member of the spiro-piperidine class of M2 proton channel inhibitors similar to BL-1743. Displays increased potency (IC50=0.92µM) compared with amantadine.
- M2 proton channel inhibitor
White solid.
Soluble in water (>25mg/ml), DMSO (>25mg/ml) or 100% ethanol (>25mg/ml).
Formel:
C10H20ClN Lagertemperatur: Raumtemperatur |
MDL:
MFCD02093801 CAS-Nummer: 1125-01-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...