Forskolin ≥99% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: (3R,4aR,5S,6S,6aS,10S,10aR,10bS)-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-3-vinyldodecahydro-1H-benzo[f]chromen-5-yl acetate

BMLCN1000010 BMLCN1000100
ENZOBMLCN1000010EA 161 EUR
ENZOBMLCN1000010 ENZOBMLCN1000100
Forskolin ≥99% (durch TLC)
Forskolin

Adenylate cyclase activator


Activates adenylate cyclase by directly interacting with the catalytic unit of the enzyme leading to an increase in the intracellular concentration of cAMP. Widely used tool for the investigation of the role of cAMP as a second messenger with a broad range of potential therapeutic applications. Inotropic agent and vasodilator. Induces platelet aggregation. Inhibits ion channels by a mechanism that does not involve cAMP. Non-competitive inhibitor of nicotinic acetylcholine receptors.


Isolated from Coleus forskohlii.

Formel: C₂₂H₃₄O₇
Molecular Weight: 410.51 g/mol
MDL: MFCD00082317
CAS-Nummer: 66575-29-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbaaa80dc33de2287e1d9a98b63daffe6e1bcb19&vid=29bdd5e4828e1ba56f5c573017c8f2b3840b6fad&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR